Anti-GRHPR Mouse Monoclonal Antibody [clone: 1A9]

Toimittaja: OriGene
ORIGCF502086EA 749 EUR
ORIGCF502086
Anti-GRHPR Mouse Monoclonal Antibody [clone: 1A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GRHPR
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR