Anti-DSTN Mouse Monoclonal Antibody [clone: 1B8]

Toimittaja: OriGene
ORIGCF502642EA 749 EUR
ORIGCF502642
Anti-DSTN Mouse Monoclonal Antibody [clone: 1B8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DSTN
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR