Anti-HDHD1A Mouse Monoclonal Antibody [clone: 6G3]

Toimittaja: OriGene
ORIGCF502644EA 749 EUR
ORIGCF502644
Anti-HDHD1A Mouse Monoclonal Antibody [clone: 6G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: HDHD1A
Clonality: Monoclonal
Clone: 6G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR