Anti-VASP Mouse Monoclonal Antibody [clone: 4D6]

Toimittaja: OriGene
ORIGCF502647EA 770 EUR
ORIGCF502647
Anti-VASP Mouse Monoclonal Antibody [clone: 4D6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR