Anti-GBA3 Mouse Monoclonal Antibody [clone: 4F10]

Toimittaja: OriGene
ORIGCF502675EA 749 EUR
ORIGCF502675
Anti-GBA3 Mouse Monoclonal Antibody [clone: 4F10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GBA3
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR