Anti-ARHGDIA Mouse Monoclonal Antibody [clone: 1F2]

Toimittaja: OriGene
ORIGCF501089EA 749 EUR
ORIGCF501089
Anti-ARHGDIA Mouse Monoclonal Antibody [clone: 1F2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: ARHGDIA
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR