Anti-KIAA1609 Mouse Monoclonal Antibody [clone: 12G10]

Toimittaja: OriGene
ORIGCF501095EA 786 EUR
ORIGCF501095
Anti-KIAA1609 Mouse Monoclonal Antibody [clone: 12G10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KIAA1609
Clonality: Monoclonal
Clone: 12G10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR