Anti-ADH1B Mouse Monoclonal Antibody [clone: 3C12]

Toimittaja: OriGene
ORIGCF502779EA 749 EUR
ORIGCF502779
Anti-ADH1B Mouse Monoclonal Antibody [clone: 3C12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ADH1B
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR