Anti-PRKAR2A Mouse Monoclonal Antibody [clone: 1A7]

Toimittaja: OriGene
ORIGCF501129EA 749 EUR
ORIGCF501129
Anti-PRKAR2A Mouse Monoclonal Antibody [clone: 1A7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PRKAR2A
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR