Anti-GSTT2 Mouse Monoclonal Antibody [clone: 2E3]

Toimittaja: OriGene
ORIGCF501829EA 770 EUR
ORIGCF501829
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 2E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:2000; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR