Anti-ANXA3 Mouse Monoclonal Antibody [clone: 5G4]

Toimittaja: OriGene
ORIGCF502139EA 770 EUR
ORIGCF502139
Anti-ANXA3 Mouse Monoclonal Antibody [clone: 5G4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: ANXA3
Clonality: Monoclonal
Clone: 5G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR