Anti-CDK2 Mouse Monoclonal Antibody [clone: 1H5]

Toimittaja: OriGene
ORIGCF502832EA 749 EUR
ORIGCF502832
Anti-CDK2 Mouse Monoclonal Antibody [clone: 1H5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CDK2
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR