Anti-SNAP25 Mouse Monoclonal Antibody [clone: 1D5]

Toimittaja: OriGene
ORIGCF502838EA 749 EUR
ORIGCF502838
Anti-SNAP25 Mouse Monoclonal Antibody [clone: 1D5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNAP25
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR