Anti-EIF1 Mouse Monoclonal Antibody [clone: 2B9]

Toimittaja: OriGene
ORIGCF502844EA 749 EUR
ORIGCF502844
Anti-EIF1 Mouse Monoclonal Antibody [clone: 2B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EIF1
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR