Anti-BSG Mouse Monoclonal Antibody [clone: 4E11]

Toimittaja: OriGene
ORIGCF501162EA 749 EUR
ORIGCF501162
Anti-BSG Mouse Monoclonal Antibody [clone: 4E11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR