Anti-BSG Mouse Monoclonal Antibody [clone: 7A7]
Toimittaja: OriGene
ORIGCF501165EA 749 EUR
ORIGCF501165
Anti-BSG Mouse Monoclonal Antibody [clone: 7A7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 7A7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human