Anti-BSG Mouse Monoclonal Antibody [clone: 9B10]

Toimittaja: OriGene
ORIGCF501188EA 749 EUR
ORIGCF501188
Anti-BSG Mouse Monoclonal Antibody [clone: 9B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR