Anti-JNK1 Mouse Monoclonal Antibody [clone: 1C2]

Toimittaja: OriGene
ORIGCF500043EA 749 EUR
ORIGCF500043
Anti-JNK1 Mouse Monoclonal Antibody [clone: 1C2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100

Type: Primary
Antigen: JNK1
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR