Anti-BFGF Mouse Monoclonal Antibody [clone: 2H11]

Toimittaja: OriGene
ORIGCF500013EA 749 EUR
ORIGCF500013
Anti-BFGF Mouse Monoclonal Antibody [clone: 2H11]
Vasta-aineet
Recommended Dilutions:Western Blot: 1:5000-10000

Type: Primary
Antigen: BFGF
Clonality: Monoclonal
Clone: 2H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR