Anti-KRT19 Mouse Monoclonal Antibody [clone: 5D1]

Toimittaja: OriGene
ORIGCF500018EA 749 EUR
ORIGCF500018
Anti-KRT19 Mouse Monoclonal Antibody [clone: 5D1]
Vasta-aineet
Recommended Dilutions:Western Blot: 1:10000-20000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: 5D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR