Anti-KRT8 Mouse Monoclonal Antibody [clone: 1B9]

Toimittaja: OriGene
ORIGCF500020EA 749 EUR
ORIGCF500020
Anti-KRT8 Mouse Monoclonal Antibody [clone: 1B9]
Vasta-aineet
Recommended Dilutions:Western Blot: 1:2500-5000; Immunofluorescence: 1:100

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: 1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR