Anti-MAPK7 Mouse Monoclonal Antibody [clone: 5B9]

Toimittaja: OriGene
ORIGCF502189EA 749 EUR
ORIGCF502189
Anti-MAPK7 Mouse Monoclonal Antibody [clone: 5B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK7
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR