Anti-TRIM2 Mouse Monoclonal Antibody [clone: 4D6]

Toimittaja: OriGene
ORIGCF502192EA 749 EUR
ORIGCF502192
Anti-TRIM2 Mouse Monoclonal Antibody [clone: 4D6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM2
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR