Anti-FAM127C Mouse Monoclonal Antibody [clone: 2D4]

Toimittaja: OriGene
ORIGCF502197EA 749 EUR
ORIGCF502197
Anti-FAM127C Mouse Monoclonal Antibody [clone: 2D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: FAM127C
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR