Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 9F3]

Toimittaja: OriGene
ORIGCF500556EA 749 EUR
ORIGCF500556
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 9F3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SLC2A5
Clonality: Monoclonal
Clone: 9F3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR