Anti-PRKY Mouse Monoclonal Antibody [clone: 2E9]

Toimittaja: OriGene
ORIGCF500561EA 749 EUR
ORIGCF500561
Anti-PRKY Mouse Monoclonal Antibody [clone: 2E9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: PRKY
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR