Anti-STAT1 Mouse Monoclonal Antibody [clone: 15H3]

Toimittaja: OriGene
ORIGCF500566EA 749 EUR
ORIGCF500566
Anti-STAT1 Mouse Monoclonal Antibody [clone: 15H3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 15H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR