Anti-CRYAB Mouse Monoclonal Antibody [clone: 6A9]

Toimittaja: OriGene
ORIGCF500581EA 749 EUR
ORIGCF500581
Anti-CRYAB Mouse Monoclonal Antibody [clone: 6A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 6A9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR