Anti-BAT1 Mouse Monoclonal Antibody [clone: 2E4]

Toimittaja: OriGene
ORIGCF501235EA 749 EUR
ORIGCF501235
Anti-BAT1 Mouse Monoclonal Antibody [clone: 2E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: BAT1
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR