Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1B6]

Toimittaja: OriGene
ORIGCF502225EA 749 EUR
ORIGCF502225
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1B6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CTDSP1
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR