Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 6A7]

Toimittaja: OriGene
ORIGCF502239EA 749 EUR
ORIGCF502239
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 6A7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 6A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR