Anti-PACSIN3 Mouse Monoclonal Antibody [clone: 4C5]

Toimittaja: OriGene
ORIGCF502240EA 770 EUR
ORIGCF502240
Anti-PACSIN3 Mouse Monoclonal Antibody [clone: 4C5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: PACSIN3
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR