Anti-CRYAB Mouse Monoclonal Antibody [clone: 1G1]

Toimittaja: OriGene
ORIGCF500590EA 749 EUR
ORIGCF500590
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1G1]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR