Anti-PPP5C Mouse Monoclonal Antibody [clone: 4G8]

Toimittaja: OriGene
ORIGCF500592EA 749 EUR
ORIGCF500592
Anti-PPP5C Mouse Monoclonal Antibody [clone: 4G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR