Anti-PPP5C Mouse Monoclonal Antibody [clone: 5G5]

Toimittaja: OriGene
ORIGCF500593EA 749 EUR
ORIGCF500593
Anti-PPP5C Mouse Monoclonal Antibody [clone: 5G5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR