Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 9G8]

Toimittaja: OriGene
ORIGCF500604EA 749 EUR
ORIGCF500604
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 9G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: SLC2A5
Clonality: Monoclonal
Clone: 9G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR