Anti-BUB1B Mouse Monoclonal Antibody [clone: 3D9]
Toimittaja: OriGene
ORIGCF500609EA 749 EUR
ORIGCF500609
Anti-BUB1B Mouse Monoclonal Antibody [clone: 3D9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: BUB1B
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human