Anti-SNX9 Mouse Monoclonal Antibody [clone: 1E2]

Toimittaja: OriGene
ORIGCF501250EA 749 EUR
ORIGCF501250
Anti-SNX9 Mouse Monoclonal Antibody [clone: 1E2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX9
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR