Anti-TACC3 Mouse Monoclonal Antibody [clone: 1E4]

Toimittaja: OriGene
ORIGCF501260EA 749 EUR
ORIGCF501260
Anti-TACC3 Mouse Monoclonal Antibody [clone: 1E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TACC3
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR