Anti-EPCAM Mouse Monoclonal Antibody [clone: 2C8]

Toimittaja: OriGene
ORIGCF500171EA 749 EUR
ORIGCF500171
Anti-EPCAM Mouse Monoclonal Antibody [clone: 2C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:50

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR