Anti-STAT3 Mouse Monoclonal Antibody [clone: 21E7]

Toimittaja: OriGene
ORIGCF500177EA 749 EUR
ORIGCF500177
Anti-STAT3 Mouse Monoclonal Antibody [clone: 21E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: STAT3
Clonality: Monoclonal
Clone: 21E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR