Anti-KRT19 Mouse Monoclonal Antibody [clone: 4E12]

Toimittaja: OriGene
ORIGCF500213EA 749 EUR
ORIGCF500213
Anti-KRT19 Mouse Monoclonal Antibody [clone: 4E12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR