Anti-ATP6V1F Mouse Monoclonal Antibody [clone: 1B8]

Toimittaja: OriGene
ORIGCF502279EA 749 EUR
ORIGCF502279
Anti-ATP6V1F Mouse Monoclonal Antibody [clone: 1B8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATP6V1F
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR