Anti-GPHN Mouse Monoclonal Antibody [clone: 7B6]

Toimittaja: OriGene
ORIGCF502284EA 749 EUR
ORIGCF502284
Anti-GPHN Mouse Monoclonal Antibody [clone: 7B6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GPHN
Clonality: Monoclonal
Clone: 7B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR