Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 4H10]

Toimittaja: OriGene
ORIGCF500632EA 749 EUR
ORIGCF500632
Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 4H10]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SLC7A8
Clonality: Monoclonal
Clone: 4H10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR