Anti-LTA4H Mouse Monoclonal Antibody [clone: 9C5]

Toimittaja: OriGene
ORIGCF500636EA 770 EUR
ORIGCF500636
Anti-LTA4H Mouse Monoclonal Antibody [clone: 9C5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: LTA4H
Clonality: Monoclonal
Clone: 9C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR