Anti-SLC2A6 Mouse Monoclonal Antibody [clone: 7E3]

Toimittaja: OriGene
ORIGCF500639EA 749 EUR
ORIGCF500639
Anti-SLC2A6 Mouse Monoclonal Antibody [clone: 7E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: SLC2A6
Clonality: Monoclonal
Clone: 7E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR