Anti-GBP2 Mouse Monoclonal Antibody [clone: 1E5]

Toimittaja: OriGene
ORIGCF500657EA 770 EUR
ORIGCF500657
Anti-GBP2 Mouse Monoclonal Antibody [clone: 1E5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR