Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 3F9]

Toimittaja: OriGene
ORIGCF500305EA 749 EUR
ORIGCF500305
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 3F9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR