Anti-PROM2 Mouse Monoclonal Antibody [clone: 5G8]

Toimittaja: OriGene
ORIGCF500315EA 770 EUR
ORIGCF500315
Anti-PROM2 Mouse Monoclonal Antibody [clone: 5G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PROM2
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR